siRNA Details
| Gene Symbol: Cntfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003929.1 | NP_001003929.1 | 3 | 291 | ||
| XM_006238055.2 | XP_006238117.1 | 2 | 213 | |||
| GenBank mRNA | S54212.1 | - | 3 | 291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntfr| Gene Symbol: Cntfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003929.1 | NP_001003929.1 | 3 | 291 | ||
| XM_006238055.2 | XP_006238117.1 | 2 | 213 | |||
| GenBank mRNA | S54212.1 | - | 3 | 291 | ||