siRNA Details
| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007001.1 | NP_001007002.1 | 15 | 1292 | ||
| XM_006238398.2 | XP_006238460.1 | 15 | 1400 | |||
| GenBank mRNA | BC083611.1 | - | 15 | 1292 | 7133514 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift74| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007001.1 | NP_001007002.1 | 15 | 1292 | ||
| XM_006238398.2 | XP_006238460.1 | 15 | 1400 | |||
| GenBank mRNA | BC083611.1 | - | 15 | 1292 | 7133514 | |