siRNA Details
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730180.4 | XP_002730226.1 | 5 | 1360 | ||
XM_003752130.3 | XP_003752178.1 | 6 | 3140 | |||
XM_006257443.2 | XP_006257505.1 | 8 | 1280 | |||
XM_006257444.2 | XP_006257506.1 | 5 | 1258 | |||
XM_006257445.2 | XP_006257507.1 | 7 | 1214 | |||
GenBank mRNA | BC104685.1 | - | 1 | 85 | ||
FQ228246.1 | - | 8 | 1242 |
