siRNA Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135873.1 | NP_001129345.1 | 9 | 1110 | ||
| XM_006257455.2 | XP_006257517.1 | 10 | 1172 | |||
| XM_008773416.1 | XP_008771638.1 | 10 | 1172 | |||
| GenBank mRNA | FQ234598.1 | - | 2 | 126 | ||

Upf3b
Genomic Map