siRNA Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015016.1 | NP_001015016.1 | 4 | 339 | ||
| GenBank mRNA | BC091196.1 | - | 4 | 339 | 7374499 | |
| FQ233803.1 | - | 4 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd72| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015016.1 | NP_001015016.1 | 4 | 339 | ||
| GenBank mRNA | BC091196.1 | - | 4 | 339 | 7374499 | |
| FQ233803.1 | - | 4 | 351 | |||