siRNA Details
| Gene Symbol: Pkdcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108007.1 | NP_001101477.1 | 3 | 617 | ||
| GenBank mRNA | BC166535.1 | - | 3 | 589 | 7191008 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkdcc| Gene Symbol: Pkdcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108007.1 | NP_001101477.1 | 3 | 617 | ||
| GenBank mRNA | BC166535.1 | - | 3 | 589 | 7191008 | |