siRNA Details
| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108083.1 | NP_001101553.1 | 3 | 1682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp44| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108083.1 | NP_001101553.1 | 3 | 1682 | ||