siRNA Details
| Gene Symbol: Jrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104612.2 | NP_001098082.1 | 2 | 1687 | ||
| XM_006241728.2 | XP_006241790.1 | 2 | 2269 | |||
| XM_008765569.1 | XP_008763791.1 | 2 | 1831 | |||
| GenBank mRNA | BC152551.1 | - | 1 | 1431 | 7133465 | |

Jrk
Genomic Map