siRNA Details
| Gene Symbol: Fam203a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007707.1 | NP_001007708.1 | 2 | 770 | ||
| GenBank mRNA | BC079158.1 | - | 2 | 770 | 7129210 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam203a| Gene Symbol: Fam203a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007707.1 | NP_001007708.1 | 2 | 770 | ||
| GenBank mRNA | BC079158.1 | - | 2 | 770 | 7129210 | |