siRNA Details
| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130695.1 | NP_001124167.1 | 22 | 2875 | ||
| XM_006242085.2 | XP_006242147.1 | 23 | 3159 | |||
| XM_008765719.1 | XP_008763941.1 | 23 | 4245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h7b| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130695.1 | NP_001124167.1 | 22 | 2875 | ||
| XM_006242085.2 | XP_006242147.1 | 23 | 3159 | |||
| XM_008765719.1 | XP_008763941.1 | 23 | 4245 | |||