siRNA Details
Gene Symbol: Tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007146.1 | NP_001007147.1 | 2 | 540 | ||
GenBank mRNA | AY724519.1 | - | 2 | 743 | ||
BC085117.1 | - | 2 | 579 | 7107186 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007146.1 | NP_001007147.1 | 2 | 540 | ||
GenBank mRNA | AY724519.1 | - | 2 | 743 | ||
BC085117.1 | - | 2 | 579 | 7107186 |