siRNA Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025703.1 | NP_001020874.1 | 5 | 549 | ||
| GenBank mRNA | BC098857.1 | - | 5 | 549 | 7380029 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Desi1| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025703.1 | NP_001020874.1 | 5 | 549 | ||
| GenBank mRNA | BC098857.1 | - | 5 | 549 | 7380029 | |