siRNA Details
| Gene Symbol: Hoxc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750414.3 | XP_003750462.1 | 2 | 967 | ||
| GenBank mRNA | S71286.1 | - | 1 | 40 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010462, AK138664, AK033186, BC053405.

Hoxc10
Genomic Map