siRNA Details
| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008880.1 | NP_001008880.1 | 3 | 416 | ||
| GenBank mRNA | AF544988.1 | - | 1 | 19 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn4b| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008880.1 | NP_001008880.1 | 3 | 416 | ||
| GenBank mRNA | AF544988.1 | - | 1 | 19 | ||