siRNA Details
| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108156.1 | NP_001101626.1 | 7 | 699 | ||
| XM_008766277.1 | XP_008764499.1 | 2 | 258 | |||
| XM_008766278.1 | XP_008764500.1 | 7 | 1095 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga11| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108156.1 | NP_001101626.1 | 7 | 699 | ||
| XM_008766277.1 | XP_008764499.1 | 2 | 258 | |||
| XM_008766278.1 | XP_008764500.1 | 7 | 1095 | |||