siRNA Details
| Gene Symbol: LOC102546312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243244.2 | XP_006243306.1 | 1 | 787 | ||
| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108156.1 | NP_001101626.1 | 18, 19 | 2405 | ||
| XM_008766277.1 | XP_008764499.1 | 13, 14 | 1943 | |||
| XM_008766278.1 | XP_008764500.1 | 18, 19 | 2780 | |||

LOC102546312
Genomic Map