siRNA Details
| Gene Symbol: Amigo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178144.1 | NP_835280.1 | 1 | 275 | ||
| GenBank mRNA | AY237731.1 | - | 1 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amigo3| Gene Symbol: Amigo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178144.1 | NP_835280.1 | 1 | 275 | ||
| GenBank mRNA | AY237731.1 | - | 1 | 275 | ||