siRNA Details
| Gene Symbol: Klc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009601.1 | NP_001009601.1 | 14 | 1898 | ||
| XM_006244516.2 | XP_006244578.1 | 14 | 1765 | |||
| XM_006244517.2 | XP_006244579.1 | 14 | 2117 | |||
| GenBank mRNA | BC087116.1 | - | 14 | 1898 | 7193915 | |

Klc4
Genomic Map