siRNA Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191809.1 | NP_001178738.1 | 9 | 2100 | ||
| XM_006245023.2 | XP_006245085.1 | 12 | 2469 | |||
| XM_006245024.2 | XP_006245086.1 | 11 | 3802 | |||
| XM_006245025.2 | XP_006245087.1 | 11 | 2365 | |||
| XM_006245026.2 | XP_006245088.1 | 12 | 2712 | |||
| XM_006245029.2 | XP_006245091.1 | 12 | 2579 | |||
| XM_006245031.2 | XP_006245093.1 | 11 | 2475 | |||
| XM_006245032.2 | XP_006245094.1 | 11 | 2608 | |||
| XM_006245033.2 | XP_006245095.1 | 11 | 2384 | |||
| XM_006245034.2 | XP_006245096.1 | 11 | 2274 | |||

Ino80d
Genomic Map