siRNA Details
Gene Symbol: Ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767164.1 | XP_008765386.1 | 8 | 582 | ||
XM_008767165.1 | XP_008765387.1 | 7 | 672 | |||
XM_008767166.1 | XP_008765388.1 | 8 | 581 | |||
XM_008767167.1 | XP_008765389.1 | 5 | 524 |
