siRNA Details
| Gene Symbol: Txndc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005559.2 | NP_001005559.2 | 2 | 1185 | ||
| NM_001146004.1 | NP_001139476.1 | 2 | 1363 | |||
| XM_006245607.2 | XP_006245669.1 | 3 | 1237 | |||
| XM_008767379.1 | XP_008765601.1 | 3 | 1760 | |||
| GenBank mRNA | BC079238.1 | - | 2 | 1363 | 7132630 | |
| BC083924.1 | - | 3 | 1243 | 7135313 | ||

Txndc2
Genomic Map