siRNA Details
| Gene Symbol: Klhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108244.1 | NP_001101714.1 | 7 | 1386 | ||
| XM_006257199.2 | XP_006257261.1 | 7 | 1797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl4| Gene Symbol: Klhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108244.1 | NP_001101714.1 | 7 | 1386 | ||
| XM_006257199.2 | XP_006257261.1 | 7 | 1797 | |||