siRNA Details
Gene Symbol: Mageb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044246.1 | NP_001037711.1 | 2 | 300 | ||
GenBank mRNA | BC097441.1 | - | 2 | 300 | 7456024 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mageb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044246.1 | NP_001037711.1 | 2 | 300 | ||
GenBank mRNA | BC097441.1 | - | 2 | 300 | 7456024 |