siRNA Details
| Gene Symbol: MGC114427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024893.1 | NP_001020064.1 | 3 | 414 | ||
| XM_008773565.1 | XP_008771787.1 | 4 | 750 | |||
| GenBank mRNA | BC097387.1 | - | 3 | 414 | 7446557 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC114427| Gene Symbol: MGC114427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024893.1 | NP_001020064.1 | 3 | 414 | ||
| XM_008773565.1 | XP_008771787.1 | 4 | 750 | |||
| GenBank mRNA | BC097387.1 | - | 3 | 414 | 7446557 | |