siRNA Details
Gene Symbol: MGC114427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024893.1 | NP_001020064.1 | 3 | 1011 | ||
XM_008773565.1 | XP_008771787.1 | 4 | 1347 | |||
GenBank mRNA | BC097387.1 | - | 3 | 1011 | 7446557 |
