siRNA Details
| Gene Symbol: Morf4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007714.1 | NP_001007715.1 | 4 | 967 | ||
| XM_006257295.1 | XP_006257357.1 | 4 | 1003 | |||
| XM_006257296.2 | XP_006257358.1 | 4 | 997 | |||
| XM_006257297.1 | XP_006257359.1 | 3 | 902 | |||
| GenBank mRNA | AY539870.1 | - | 1 | 682 | ||
| BC083606.1 | - | 4 | 967 | 7133330 | ||
| FQ212552.1 | - | 3 | 865 | |||
| FQ220931.1 | - | 1 | 134 | |||
| FQ232538.1 | - | 4 | 966 | |||

Morf4l2
Genomic Map