siRNA Details
| Gene Symbol: Tnfsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181384.2 | NP_852049.1 | 3 | 837 | ||
| GenBank mRNA | AY259541.1 | - | 3 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf9| Gene Symbol: Tnfsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181384.2 | NP_852049.1 | 3 | 837 | ||
| GenBank mRNA | AY259541.1 | - | 3 | 837 | ||