siRNA Details
| Gene Symbol: Lhfpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181387.2 | NP_852052.1 | 2 | 479 | ||
| GenBank mRNA | AY278321.1 | - | 2 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhfpl4| Gene Symbol: Lhfpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181387.2 | NP_852052.1 | 2 | 479 | ||
| GenBank mRNA | AY278321.1 | - | 2 | 479 | ||