siRNA Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182675.1 | NP_872616.1 | 3 | 225 | ||
| XM_006245981.2 | XP_006246043.2 | 4 | 322 | |||
| GenBank mRNA | AY190240.1 | - | 3 | 225 | ||
| BC128720.1 | - | 3 | 266 | 8374477 | ||

Rab40c
Genomic Map