siRNA Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277232.1 | NP_001264161.1 | 9 | 856 | ||
| NM_182953.2 | NP_891998.1 | 9 | 908 | |||
| XM_006234100.1 | XP_006234162.1 | 9 | 931 | |||
| GenBank mRNA | AY334013.1 | - | 8 | 805 | ||
| BC160820.1 | - | 8 | 932 | 7459041 | ||

Nek6
Genomic Map