siRNA Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007721.1 | NP_001007722.1 | 3 | 305 | ||
| GenBank mRNA | BC081865.1 | - | 3 | 305 | 7096470 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emp2| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007721.1 | NP_001007722.1 | 3 | 305 | ||
| GenBank mRNA | BC081865.1 | - | 3 | 305 | 7096470 | |