siRNA Details
| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108264.1 | NP_001101734.1 | 12, 13 | 1315 | ||
| XM_006245802.2 | XP_006245864.1 | 12, 13 | 1293 | |||
| XM_008767491.1 | XP_008765713.1 | 12, 13 | 1099 | |||
| XM_008767492.1 | XP_008765714.1 | 11, 12 | 2157 | |||

Alg1
Genomic Map