siRNA Details
| Gene Symbol: Sat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108278.1 | NP_001101748.1 | 4 | 406 | ||
| XM_006246743.1 | XP_006246805.1 | 5 | 527 | |||
| XM_006246744.2 | XP_006246806.1 | 5 | 428 | |||
| XM_008767837.1 | XP_008766059.1 | 4 | 443 | |||
| GenBank mRNA | FQ218736.1 | - | 5 | 387 | ||

Sat2
Genomic Map