siRNA Details
| Gene Symbol: Tnfsf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001513.2 | NP_001001513.2 | 7 | 876 | ||
| XM_006246745.2 | XP_006246807.1 | 7 | 1119 | |||
| GenBank mRNA | AY607588.1 | - | 7 | 583 | ||
| BC079107.1 | - | 7 | 876 | 7125293 | ||

Tnfsf12
Genomic Map