siRNA Details
Gene Symbol: Tusc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039163.1 | NP_001034252.1 | 1 | 604 | ||
GenBank mRNA | AB218813.1 | - | 1 | 604 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tusc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039163.1 | NP_001034252.1 | 1 | 604 | ||
GenBank mRNA | AB218813.1 | - | 1 | 604 |