siRNA Details
| Gene Symbol: Ankrd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134699.1 | NP_001128171.1 | 4 | 990 | ||
| GenBank mRNA | BC166790.1 | - | 4 | 990 | 7936453 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd40| Gene Symbol: Ankrd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134699.1 | NP_001128171.1 | 4 | 990 | ||
| GenBank mRNA | BC166790.1 | - | 4 | 990 | 7936453 | |