siRNA Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108297.1 | NP_001101767.1 | 4, 5 | 653 | ||
| XM_006247347.1 | XP_006247409.1 | 4, 5 | 688 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdma| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108297.1 | NP_001101767.1 | 4, 5 | 653 | ||
| XM_006247347.1 | XP_006247409.1 | 4, 5 | 688 | |||