siRNA Details
| Gene Symbol: Cdr2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247771.2 | XP_006247833.1 | 5 | 1123 | ||
| XM_006247772.1 | XP_006247834.1 | 5 | 1025 | |||
| XM_008768410.1 | XP_008766632.1 | 5 | 845 | |||
| XM_340933.7 | XP_340934.5 | 5 | 1127 | |||

Cdr2l
Genomic Map