siRNA Details
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012472.1 | NP_001012490.1 | 9 | 1087 | ||
XM_006248635.2 | XP_006248697.1 | 9 | 1132 | |||
XM_006248636.2 | XP_006248698.1 | 8 | 1027 | |||
GenBank mRNA | BC089898.1 | - | 9 | 1087 | 7382217 |
