siRNA Details
| Gene Symbol: Ska3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108379.1 | NP_001101849.1 | 8 | 1452 | ||
| XM_006252074.1 | XP_006252136.1 | 8 | 1258 | |||
| XM_006252075.1 | XP_006252137.1 | 8 | 1258 | |||
| XM_008770758.1 | XP_008768980.1 | 8 | 1255 | |||
| GenBank mRNA | BC166461.1 | - | 8 | 1229 | 9030107 | |

Ska3
Genomic Map