siRNA Details
Gene Symbol: Micu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134396.2 | NP_599223.2 | 11 | 1083 | ||
GenBank mRNA | EX491796.1 | - | 2 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Micu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134396.2 | NP_599223.2 | 11 | 1083 | ||
GenBank mRNA | EX491796.1 | - | 2 | 98 |