siRNA Details
| Gene Symbol: Micu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134396.2 | NP_599223.2 | 10 | 1013 | ||
| GenBank mRNA | DY317464.1 | - | 8 | 663 | ||
| EX491796.1 | - | 1 | 28 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Micu2| Gene Symbol: Micu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134396.2 | NP_599223.2 | 10 | 1013 | ||
| GenBank mRNA | DY317464.1 | - | 8 | 663 | ||
| EX491796.1 | - | 1 | 28 | |||