siRNA Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013188.2 | NP_001013206.1 | 3 | 261 | ||
| GenBank mRNA | BC058504.1 | - | 3 | 265 | 6887991 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Leprotl1| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013188.2 | NP_001013206.1 | 3 | 261 | ||
| GenBank mRNA | BC058504.1 | - | 3 | 265 | 6887991 | |