siRNA Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728416.4 | XP_002728462.1 | 2, 3 | 379 | ||
| GenBank mRNA | FQ214349.1 | - | 2, 3 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim19| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728416.4 | XP_002728462.1 | 2, 3 | 379 | ||
| GenBank mRNA | FQ214349.1 | - | 2, 3 | 350 | ||