siRNA Details
Gene Symbol: Cd83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108410.1 | NP_001101880.1 | 3 | 418 | ||
GenBank mRNA | BC166582.1 | - | 3 | 329 | 7385133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108410.1 | NP_001101880.1 | 3 | 418 | ||
GenBank mRNA | BC166582.1 | - | 3 | 329 | 7385133 |