siRNA Details
| Gene Symbol: Pxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025719.1 | NP_001020890.1 | 3 | 603 | ||
| XM_006253841.2 | XP_006253903.1 | 3 | 1255 | |||
| GenBank mRNA | BC099145.1 | - | 3 | 603 | 7325668 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pxdc1| Gene Symbol: Pxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025719.1 | NP_001020890.1 | 3 | 603 | ||
| XM_006253841.2 | XP_006253903.1 | 3 | 1255 | |||
| GenBank mRNA | BC099145.1 | - | 3 | 603 | 7325668 | |