siRNA Details
| Gene Symbol: Bphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037206.1 | NP_001032283.1 | 4, 5 | 544 | ||
| GenBank mRNA | BC105908.1 | - | 4, 5 | 544 | 7388526 | |
| FQ215116.1 | - | 4, 5 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bphl| Gene Symbol: Bphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037206.1 | NP_001032283.1 | 4, 5 | 544 | ||
| GenBank mRNA | BC105908.1 | - | 4, 5 | 544 | 7388526 | |
| FQ215116.1 | - | 4, 5 | 554 | |||