siRNA Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108460.1 | NP_001101930.1 | 2 | 180 | ||
| XM_008758672.1 | XP_008756894.1 | 2 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hddc2| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108460.1 | NP_001101930.1 | 2 | 180 | ||
| XM_008758672.1 | XP_008756894.1 | 2 | 201 | |||