siRNA Details
| Gene Symbol: Zfp52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014158.1 | NP_001014180.1 | 6 | 2166 | ||
| XM_006228003.2 | XP_006228065.1 | 5 | 1899 | |||
| GenBank mRNA | BC085827.1 | - | 6 | 2166 | 7131994 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp52| Gene Symbol: Zfp52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014158.1 | NP_001014180.1 | 6 | 2166 | ||
| XM_006228003.2 | XP_006228065.1 | 5 | 1899 | |||
| GenBank mRNA | BC085827.1 | - | 6 | 2166 | 7131994 | |