siRNA Details
| Gene Symbol: Meis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108472.1 | NP_001101942.1 | 7 | 714 | ||
| XM_006228346.2 | XP_006228408.1 | 5 | 928 | |||
| XM_006228347.2 | XP_006228409.1 | 6 | 1461 | |||
| XM_006228348.2 | XP_006228410.1 | 5 | 928 | |||
| XM_006228349.2 | XP_006228411.1 | 6 | 775 | |||
| XM_006228350.2 | XP_006228412.1 | 6 | 724 | |||
| XM_006228351.2 | XP_006228413.1 | 6 | 724 | |||
| XM_008758950.1 | XP_008757172.1 | 6 | 775 | |||
| GenBank mRNA | BC160854.1 | - | 5 | 828 | 8364874 | |

Meis3
Genomic Map